missing translation for 'onlineSavingsMsg'
Learn More

Thianthrene 98.0+%, TCI America™

Catalog No. T01845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01845G 5 g
T018425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01845G Supplier TCI America Supplier No. T01845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-85-3
Molecular Formula C12H8S2
Molecular Weight (g/mol) 216.32
MDL Number MFCD00005065
InChI Key GVIJJXMXTUZIOD-UHFFFAOYSA-N
PubChem CID 7109
ChEBI CHEBI:64511
IUPAC Name thianthrene
SMILES S1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thianthrene