missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamphenicol 98.0+%, TCI America™
Supplier: TCI America T280225G
Specifications
| Thiamphenicol | |
| 166°C | |
| C12H15Cl2NO5S | |
| 25 g | |
| OTVAEFIXJLOWRX-NXEZZACHSA-N | |
| 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide | |
| 27200 | |
| 356.21 | |
| Crystalline Powder |
| 15318-45-3 | |
| White | |
| MFCD00467983 | |
| thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O | |
| 356.214 | |
| CHEBI:32215 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 15318-45-3 | |
| 356.214 | |
| OTVAEFIXJLOWRX-NXEZZACHSA-N | |
| 27200 | |
| 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
| C12H15Cl2NO5S | |
| MFCD00467983 | |
| thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol | |
| CHEBI:32215 | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O |
Safety and Handling
EINECSNumber : (9)-0816
RTECSNumber : AB6680000
TSCA : No