missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol 98.0+%, TCI America™

Catalog No. p-7137427
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28025G 5 g
T280225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28025G Supplier TCI America Supplier No. T28025G
Only null left

Chemical Identifiers

CAS 15318-45-3
Molecular Formula C12H15Cl2NO5S
Molecular Weight (g/mol) 356.214
MDL Number MFCD00467983
InChI Key OTVAEFIXJLOWRX-NXEZZACHSA-N
Synonym thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol
PubChem CID 27200
ChEBI CHEBI:32215
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O

Specifications

Melting Point 166°C
Color White
Quantity 5 g
Formula Weight 356.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Thiamphenicol
EINECSNumber (9)-0816
RTECSNumber AB6680000
TSCA No