missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18607360
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607360 25 kg Amber Glass Bottle
18607356 25 g Amber Glass Bottle
18607357 100 g Amber Glass Bottle
18607358 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18607360 Supplier Spectrum Chemical Mfg Cor Supplier No. TH11625KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TH116, 67-03-8, C12H17ClN4OS·HCl

Spectrum™ Chemical Thiamine Hydrochloride, USP is the hydrochloride salt of vitamin B1 and used to treat or prevent vitamin B1 deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.26
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
IUPAC Name hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride
SMILES [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N

Specifications

CAS Max % 1
pH 2.7 to 3.4
Ignition Residue 0.002
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Thiamine Hydrochloride