missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™

Catalog No. 02004477
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004477 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02004477 Supplier Avantor Supplier No. 411006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.263
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

Packaging Poly Pail
Quantity 2.5 kg
Formula Weight 337.27
Grade FCC/USP
Chemical Name or Material Thiamine Hydrochloride