missing translation for 'onlineSavingsMsg'
Learn More

Thiamine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015125
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015125 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015125 Supplier Merck Emd Millipore Supplier No. PHR10371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Specifications

CAS 67-03-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C12H17 ClN4 OS · HCl
MDL Number MFCD00012780
Melting Point 250°C
Synonym Aneurine hydrochloride; Vitamin B1 hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 337.27
Grade Certified Reference Material
Show More Show Less