missing translation for 'onlineSavingsMsg'
Learn More

Thiacetazone 98.0+%, TCI America™

Catalog No. T3632250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3632250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3632250MG Supplier TCI America Supplier No. T3632250MG

Chemical Identifiers

CAS 104-06-3
Molecular Formula C10H12N4OS
Molecular Weight (g/mol) 236.293
MDL Number MFCD00022157
InChI Key SRVJKTDHMYAMHA-WUXMJOGZSA-N
Synonym thioacetazone, thiacetazone, amithiozone, ambathizon, benzothiozane, conteben, benzothiozon, benthiozone, parazone, thioacetazon
PubChem CID 9568512
IUPAC Name N-[4-[(E)-(carbamothioylhydrazinylidene)methyl]phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C=NNC(=S)N

Specifications

Melting Point 223°C
Quantity 250 mg
Formula Weight 236.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Thiacetazone
RTECSNumber AE3850000
TSCA No