missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers

Catalog No. 03528
Change view
Click to view available options
Capacity:
3 or 5 mL
3, 5, 7 or 10 mL
5, 7 or 10 mL
7 to 10 mL
7, 10, 15 or 25 mL
Quantity:
3 Tubes
4 Tubes
5 Tubes
8 Tubes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Quantity
03528 3, 5, 7 or 10 mL 3 Tubes
0353035 7, 10, 15 or 25 mL 5 Tubes
035305 5, 7 or 10 mL 8 Tubes
0353120 3 or 5 mL 5 Tubes
03600165 7 to 10 mL 4 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03528 Supplier Sonoco™ Supplier No. TS364
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps

  • Two-piece assembly with snap-lock fasteners holds contents in place
  • Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples.

†Includes slotted area for up to four slides.

Specifications

Quantity 3 Tubes
Applications Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples
Dimensions (LxWxH) 105 x 17 mm
Capacity 3, 5, 7 or 10 mL
Material Polystyrene Foam