Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Solid Discs
Suitable for conveniently introducing the sample without causing the system to leak. Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
Supplier:  MilliporeSigma™ Supelco™ 23159
 Afficher plus de versions de ce produit
Numéro de catalogue. 111020861
Il en reste null
Description
Description
  • An improved version over the original Thermogreen™ LB-1 Septa
  • Extremely low bleed over a wide range of inlet temperatures (100° to 350°C)
  • Easy needle penetration
  • High puncture tolerance (ideal for autosamplers)
  • Rubber formulation exclusive to Supelco™
  • Pre-conditioned, ready to use
Spécifications
Identifiants chimiques
Green
Solid Disc
Thermogreen
Rubber
100°C to 350°C
Septa, LB-2
Spécifications
21/32 in.
17 mm
50/Pk.
100°C to 350°C
Finnigan TRACE™, Fisons 8000 Series
Green
Rubber
Solid Disc
Thermogreen
Septa, LB-2
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC