missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Shimadzu Plug

Suitable for conveniently introducing the sample without causing the system to leak. Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.

$81.46 - $280.57

Specifications

Diameter (Metric) Outer 6 mm
Style Shimadzu Plug
Product Line Thermogreen
For Use With (Equipment) Shimadzu Gas Chromatographs
Height (Metric) 9 mm
View More Specs

Products 2
Catalog Number Mfr. No. No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. No. per Pack Price Quantity & Availability  
111011626
View Documents Promotion Details
MilliporeSigma™ Supelco™
20608
10/Pk.
Each for $81.46
Only null left
 
111020840
View Documents Promotion Details
MilliporeSigma™ Supelco™
20633
50/Pk.
Each for $280.57
Only null left
 
Description

Description

For use with:

  • Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
  • Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
  • Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
  • Influent sampling ports in gas chromatographs
Specifications

Specifications

6 mm
Thermogreen
9 mm
Shimadzu Plug
Shimadzu Gas Chromatographs
Septa, LB-2
Videos
SDS
Documents

Documents

Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC