missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Consul-Mount™

Catalog No. 9990441
Change view
Click to view available options
Product Type:
Cytology Media
Mountant
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
9990441 Cytology Media
9990440 Mountant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9990441 Supplier Epredia™ Supplier No. 9990441
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed specifically for use with the Epredia™ Consul™ coverslipper

  • Permanent, organic mountant
  • Xylene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Medium (125-200 cps at 24°C)

Specifications

Color Clear
Product Type Cytology Media
Quantity 500 mL
For Use With (Application) For use with Epredia™ Consul™ coverslipper
Form Liquid