Learn More
Thermo Scientific™ Samco™ Graduated Transfer Pipettes
GREENER_CHOICE
Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.
Numéro de catalogue. 02708011
Il en reste null
Description
Description
Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Spécifications
Spécifications
1.8 mL
7.1 mm
Sterile
Yes
Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs
15.2 cm
Yes
Transfer Pipette, Graduated to .5mL, 1mL, 1.5mL, 2mL
LDPE
25
0.5, 1, 1.5, 2 mL
4.6 mL
Graduated Transfer Pipette
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS