missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ CTA w/1% Lactose
Specifications
Specifications
| Additive | 1% Lactose |
| Media Type | CTA Agar (Cystine Trypticase Agar) |
| Product Type | Microbiology Media |
| For Use With (Application) | Determination of fermentation reactions by fastidious microorganisms such as &italic_begin;Neisseria&italic_end;, pneumococci, streptococci, and nonspore forming anaerobes |
| No. per Pack | 20 |
| Quantity | 4 mL |