missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peroxide Assay Reagent A

Catalog No. PI23281
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23281 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23281 Supplier Thermo Scientific™ Supplier No. LSG23281

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Measure total peroxide levels in either aqueous or lipid-containing laboratory reagents

Thermo Scientific Pierce Peroxide Assay Reagent A is a formulation consisting of 25mM ammonium ferrous sulfate. This formulation is one of the component of Quantitative Peroxide Assay Kit used for detecting and measuring hydrogen peroxide levels based on the oxidation of ferrous to ferric ion in the presence of xylenol orange, present in the biological and other liquid samples.

Applications of Pierce Quantitative Peroxide Assay Kits:

  • General hydrogen peroxide (H2O2) measurement in protein samples
  • Quantitation of peroxides in detergents
  • Assessment of the effects of peroxides on cellular activity
  • Determination of protein glycation; Measurement of peroxide accumulation in lipids

In these assay kits, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With the aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In the lipid compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the know extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution. Most proteins do not interfere with these assays, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.

Related Products

Pierce™ Quantitative Peroxide Assay Kit (Aqueous) (Cat. No. 23280)

Pierce™ Quantitative Peroxide Assay Kit (Lipid) (Cat. No. 23285)

Pierce™ Peroxide Assay Reagent B (Cat. No. 23282)

Pierce™ Peroxide Assay Reagent C (Cat. No. 23283)

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Assay Peroxide Assay Reagent A
Detection Method Absorbance
Product Line Pierce
Product Type Quantitative Peroxide Assay
Specificity Not Target-Specific
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 1 mL

For Research Use Only. Not for use in diagnostic procedures.