missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HRP-Conjugated Streptavidin

Catalog No. ENN100
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENN100 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENN100 Supplier Thermo Scientific™ Supplier No. N100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin from Streptomyces avidinii conjugated to highly purified horseradish peroxidase (HRP)

  • Validated for use with Matched Antibody Pairs and MiniKits
  • Pre-defined working ranges save time and facilitate assay optimization

Specifications

Concentration 1.25 mg/mL
Conjugate HRP
Quantity 0.25 mL
Content And Storage Store between 2°C and 8°C.
Form Liquid
Product Type Streptavidin Conjugate
Target Molecule Biotin
Label or Dye HRP (Horseradish Peroxidase)
In which solvent is the HRP-Conjugated Streptavidin (Cat. No. N100) dissolved?

The HRP-Conjugated Streptavidin (Cat. No. N100) is dissolved in an aqueous solution of 0.99% BSA and 0.1% ProCelin preservative.

For Research Use Only. Not for use in diagnostic procedures.