missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt

Catalog No. PI88293
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88293 100 mg
PI88294 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88293 Supplier Thermo Scientific™ Supplier No. 88293
  • ONLINE EXCLUSIVE PRICE
    $304.79
    $166.46 / Each

    Save $138.33

    45.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.

Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.

Thermo Scientific Pierce D-Luciferin is formulated at greater than 99% purity as both monosodium and monopotassium salts to provide convenience and high performance at a low cost in reporter assays involving firefly luciferase.

Luciferin is the substrate for bioluminescence reactions with firefly luciferase enzyme. Lyophilized potassium and sodium salts of firefly luciferin are stable and easily soluble, and both formulations are compatible with a wide variety of luciferase-based in vivo imaging and in vitro bioluminescent assays. (The decision to use one or the other form is a matter of researcher preference.) Applications include ATP assays, gene reporter assays, high-throughput sequencing and various contamination assays throughout the biotechnology field. For in vivo imaging, the injection of luciferin in model systems enables real-time, non-invasive monitoring of disease progression and/or drug efficacy through bioluminescence.

Highlights:

  • High purity, typically greater than 99.7%, as verified by HPLC and FTIR
  • Great performance with in vitro applications
  • Suitable for in vivo imaging
Applications:
  • Gene reporter assays
  • ATP assays
  • High throughput sequencing
  • Contamination assays
  • Bioluminescent in vivo imaging
  • Drug screening
  • Multiplexed assays to study off-target effects
  • Signal transduction pathway analysis

Specifications

Content And Storage Store at or below -20°C.
Detection Method Bioluminescence
For Use With (Application) ATP Assay, Gene Reporter Assay
Product Type D-Luciferin, Monopotassium Salt
Product Line Pierce
Quantity 100 mg
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate

For Research Use Only. Not for use in diagnostic procedures.