Learn More
Thermo Scientific™ Collection Plates (Polypropylene) for Pierce™ 96-Well Filter Spin Plates
Catalog No. PI89813
Change view
Click to view available options
Quantity:
6 plates
Catalog No. Quantity
PI89813 6 plates
1 options
Catalog No. PI89813 Supplier Thermo Scientific™ Supplier No. 89813
Collection plates and balance plates for use with 96-well filter spin plates
These Thermo Scientific Plate Accessories are collection plates and centrifuge balance plates for affinity purification and desalting resin products that use Thermo Scientific Pierce 96-Well Filter Spin Plates.
Related Products
Balance Plate for Pierce™ 96-Well Filter Spin Plates
Specifications
Content And Storage 0.0
Product Line Pierce
Product Type Collection Plate
Material Polypropylene
No. of Wells 96
Plate Compatibility 96-Well Plates
Quantity 6 plates
For Research Use Only. Not for use in diagnostic procedures.