missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-HA Agarose

Catalog No. PI26182
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26182 5 mL
PI26181 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI26182 Supplier Thermo Scientific™ Supplier No. 26182

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation

An HA tag is frequently incorporated into recombinant proteins for a variety of purposes. An anti-HA antibody can then be used to detect the protein when doing studies with transfected cells.
Host Species: Mouse
Cross Reactivity: Tag
Isotype: IgG1
Clone: 2-2.2.14
Immunogen: HA peptide YPYDVPDYA derivitized to ovalbumin
Formulation: 10mL Purified Antibody in PBS, with 0.05% sodium azide; pH7.2
Synonyms: human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
26182 is supplied as 10mL of antibody on crosslinked 4% beaded agarose supplied as a 50% slurry in 0.1M phosphate, 0.15M NaCl, pH 7.2 with 0.05% sodium azide. Specific for the HA peptide YPYDVPDYA. The anti-HA agarose has been used successfully in immunoprecipitation applications. The anti-HA agarose has 3.5mg mouse anti-HA IgG1 monoclonal antibody conjugated permL of settled agarose resin. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.

Specifications

Product Type Agarose
Column Type Agarose Resin, Affinity
Format Bottle
Conjugate HA-epitope tag (YPYDVPDYA)
Purification Target Anti-HA Antibody
Content And Storage 5 mL of settle resin, provided as 10 mL of 50% slurry. Store at 2–8°C.
Quantity 5 mL
What type of recombinant protein purification resins do you offer?

We offer a variety of resins targeting His-tagged fusion proteins or GST fusion proteins. Please see this selection table to choose the right resin for your experiments. Table available at http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/recombinant-protein-purification.html

We also offer resins (both agarose and magnetic bead-based) with immobilized anti-c-Myc and anti-HA antibodies for targeting c-Myc and HA-tagged proteins respectively. Please check out the following links for the specific products:

-Anti-c-Myc agarose (https://www.thermofisher.com/order/catalog/product/20169?ICID=search-product)
-Anti-c Myc magnetic beads (https://www.thermofisher.com/order/catalog/product/88843?ICID=search-product)
-Anti-HA agarose (https://www.thermofisher.com/order/genome-database/antibody/HA-Epitope-Tag-Antibody-clone-2-2-2-14-Monoclonal/26181)
-Anti-HA magnetic beads: (https://www.thermofisher.com/order/catalog/product/88837?ICID=search-product)

For Research Use Only. Not for use in diagnostic procedures.