missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Luminaris HiGreen qPCR Master Mix, fluorescein
Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.
Supplier: Thermo Scientific™ K0982
Includes: Luminaris HiGreen Fluorescein qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer. The master mix contains fluorescein passive reference dye for specific real-time platfo
Description
- UDG in the master mix to prevent carry-over contamination
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX or fluorescein for different real-time platforms
- Standard cycling protocols
- Excellent qPCR performance
- Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity: single copy detection
- Wide linear range: accurate quantification across 9 orders of magnitude
- Reproducibility and convenience: ready-to-use 2X master mix
Specifications
| 2X | |
| SYBR | |
| qPCR | |
| Standard | |
| Luminaris | |
| 500 Reactions | |
| 2 X | |
| DNA, dsDNA |
| Contains: • 4 x 1.25 mL Luminaris HiGreen Fluorescein qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 500 x 20 μL reactions. The master mix contains fluorescein passive reference dye for specific real-time platforms. • 4 x 1.25 mL Nuclease-Free Water Store at -20°C. |
|
| High | |
| Taq DNA Polymerase | |
| BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
| qPCR Master Mix | |
| Gene Expression | |
| 500 Reactions |