missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Every Lab Package
Description
The Everylab Microcentrifuge and Liquid Handling package provides a convenient offering that includes a microcentrifuge, Finnpipettes F2 GLP Kit (includes 4 single channel pipettes, stand, and tips), and a case of 1.5 mL microcentrifuge tubes (5000 tubes).
Covers basic and complex workflows
- Our ventilated and refrigerated microcentrifuges reduce spin times by up to 20%, making them ideal for high-end molecular or genomic research applications
- The Thermo Scientific™ included 24 X 1.5/2.0mL rotor with ClickSeal™ biocontainment lid allows you to run tubes from 1.5 to 2.0mL tube to mini-preps and spin columns
- Pipettes cover a range of volumes to match necessary volumes, from 0.2-2μL to 100-1000μL
- Snap Cap tubes offer versatility in the lab: Can be frozen or heated in a water bath
Save time with faster sample processing
- Lightweight rotors provide acceleration up to 21,100 x g in 12 seconds.
- Refrigerated models offer cooling from room temperature to set temperature in just 9 minutes.
- Color coding of pipettes helps to select the correct pipette volume while working
Ideal for everyday laboratory use
- High performance engineered polymer rotors are designed for outstanding chemical resistance to minimize corrosion from aggressive reagents
- Conforms to the latest clinical and safety standards including 98/79/EC In Vitro Diagnostic Directive
- Pipettes are fully autoclavable, minimizing disruption and downtime
- Finnpipette pipettes are made of polyvinylidene fluoride (PVDF) components that stand up to harsh chemical, UV light and physical damage
Specifications
Specifications
| Capacity | 24 x 1.5/2mL |
| Max. Speed | 14,800 rpm |
| Max. RCF | 21,100 x g |
| Includes |
|
| Refrigerated | No |
| Product Line | Sorvall |
| Voltage | 120 V |
| Product Type | Microcentrifuge |