Learn More
Thermo Scientific™ BetaSil™ C18 Guard Cartridges
Economically extends HPLC column lifetime
Supplier:  Thermo Scientific™ 70105012101
 View more versions of this product
Catalog No. 70105012101
Only null left
Explore available promotions
Description
Description
Economically extends HPLC column lifetime
Features of Betasil C18 Solid Phase
  • Thermo Scientific Chemistry-on 100Å silica
  • High surface area with high bonded phase coverage
Specifications
Specifications
5 μm
2.1 mm
BetaSil
10 mm
C18
Product Suggestions
Product Suggestions