Learn More
Thermo Scientific™ Cap-It Tool (for domed cap strips)
Catalog No. AB0536
Change view
Click to view available options
Quantity:
1 unit
Catalog No. Quantity
AB0536 1 unit
1 options
Catalog No. AB0536 Supplier Thermo Scientific™ Supplier No. AB0536
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Make sealing PCR and qPCR plates or tube strips easy
Thermo Scientific Sealing Accessories feature the Cap-IT Tool, which helps you add domed caps to tube strips and plates and helps seal multiple tubes at the same time. The Adhesive Seal Applicator helps you effectively seal PCR and qPCR plates and storage plates.
Highlights
Cap-It Tool
  • Designed to assist in applying domed caps to tube strips and plates
  • It minimizes the pressure required to seal the caps effectively
  • Enables multiple tubes to be sealed simultaneously
Adhesive Seal Applicator
  • Helps ensure effective and consistent sealing of PCR/qPCR plates or storage plates with any adhesive seal
Hazardous:No
Storage Condition: Ambient
Specifications
Compatibility 8-Well strips, 96-Well Plates, 96-Well Plate, Fast (0.1 ml), 48-Well Plates, 24-Well Plates
Quantity 1 unit