Learn More
Alkali Scientific ThermaSeal™ Polypropylene Sealing Film, Sterile, 100/pk
Numéro de catalogue. 502389246
Encompass
Only recommended for structurally rigid polypropylene PCR plates that dont warp. A less rigid polypropylene film that minimizes aerosols upon removal. Certified DNase- & RNase- free. Features good chemical resistance.
Numéro de catalogue. 502389246 Fournisseur Alkali Scientific Code fournisseur SF30MLS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Only recommended for structurally rigid polypropylene PCR plates that dont warp. A less rigid polypropylene film that minimizes aerosols upon removal. Certified DNase- & RNase- free. Features good chemical resistance.