missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Thermal Printer Paper for Clinical Chemistry Analyzers

Numéro de catalogue. 361016854
Change view
Click to view available options
Description:
82 ft. x 110 mm wide, 47 mm dia.
82 ft. x 80 mm wide, 47 mm dia.
Quantity:
10 Rolls/Box; 48 Rolls/Case
5 Rolls/Box; 24 Rolls/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Description Quantity
361016854 82 ft. x 80 mm wide, 47 mm dia. 10 Rolls/Box; 48 Rolls/Case
361016855 82 ft. x 110 mm wide, 47 mm dia. 5 Rolls/Box; 24 Rolls/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 361016854
Il en reste null

Thermal printer paper for use with popular chemistry analyzers.

  • For use with Roche™ Cobas Mira™ analyzer, Tosoh™ 360 analyzers
  • Size (L x W): 82 ft. x 80mm
  • Dia.: 47mm
  • Quantity: 10 rolls/box; 48 rolls/case

Spécifications

Type Printer Paper
Description 82 ft. x 80 mm wide, 47 mm dia.
For Use With (Equipment) Roche Cobas Mira analyzer, Tosoh 360 analyzers
Quantity 10 Rolls/Box; 48 Rolls/Case