Learn More
Lonza Walkersville Inc TheraPEAK X-VIVO-10 Serum-free Hematopoietic Cell Medium
Numéro de catalogue. NC2226656
Encompass
TheraPEAK X-VIVO 10 Media formulations are designed to support slower growing, less mature cells such as hematopoetic stem cells and natural killer cells.TheraPEAK X-VIVO 10 also supports the generation of Lymphokine activated killer (LAK) cells and peripheral blood lymphocytes (PBL). TheraPEAKX-VIVO 10 Media is available as a 1X liquid in 3 convenient formulations as off the shelf catalog items.Additional customization of packaging available.Applications:Proliferation of peripheral blood lymphocytesProliferation of tumor infiltrating lymphocytesCryopreservation of human tissueCultivation of human monocytes and macrophagesCultivation of stem cellsCultivation of dendritic cellsCultivation of CAR-T cells
Numéro de catalogue. NC2226656 Fournisseur Lonza Walkersville Inc Code fournisseur BEBP02055Q
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
TheraPEAK X-VIVO 10 Media formulations are designed to support slower growing, less mature cells such as hematopoetic stem cells and natural killer cells.TheraPEAK X-VIVO 10 also supports the generation of Lymphokine activated killer (LAK) cells and peripheral blood lymphocytes (PBL). TheraPEAKX-VIVO 10 Media is available as a 1X liquid in 3 convenient formulations as off the shelf catalog items.Additional customization of packaging available.Applications:Proliferation of peripheral blood lymphocytesProliferation of tumor infiltrating lymphocytesCryopreservation of human tissueCultivation of human monocytes and macrophagesCultivation of stem cellsCultivation of dendritic cellsCultivation of CAR-T cells