missing translation for 'onlineSavingsMsg'
Learn More

Theophylline Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019636
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019636 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019636 Supplier Merck Emd Millipore Supplier No. PHR10231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Specifications

CAS 58-55-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C7H8N4O2
MDL Number MFCD00079619
Synonym 1,3-Dimethylxanthine; 2,6-Dihydroxy-1,3-dimethylpurine; 3,7-Dihydro-1,3-dimethyl-1 H-purine-2,6-dione
Type Certified Reference Material
Quantity 1 g
Formula Weight 180.16
Grade Certified Reference Material