missing translation for 'onlineSavingsMsg'
Learn More

Theobromine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015652
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015652 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015652 Supplier Merck Emd Millipore Supplier No. PHR1837100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 83-67-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H8N4O2
MDL Number MFCD00022830
Melting Point 345°C to 350°C
Synonym Theobromine; 2,6-Dihydroxy-3,7-dimethylpurine; 3,7-Dihydro-3,7-dimethyl-1 H-purine-2,6-dione; 3,7-Dimethylxanthine
Type Certified Reference Material
Quantity 100 mg
Formula Weight 180.16
Grade Certified Reference Material