Learn More
SOMNI Scientific™ The SOMNI Breeze
Catalog No. 04777173
Encompass
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04777173 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04777173 Supplier SOMNI Scientific™ Supplier No. PH4017
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Attaches to a patient warming device to dry up to 3 breathing circuits at a time
  • Attaches to a patient warming device to dry up to 3 breathing circuits at a time
  • Easy installation
  • Dry up to 3 breathing circuits at the same time
  • Fits Bair Hugger, Equator and Desert Breeze patient warmers
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) Bair Hugger, Equator and Desert Breeze Patient Warmers