missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thaumatin, TCI America™
Spécifications
Spécifications
| Quantity | 1 g |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Thaumatin |
Sécurité et manipulation
| TSCA | No |