missing translation for 'onlineSavingsMsg'
Learn More

Thallium ICP Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PTL1KN500

Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. PTL1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7440-28-0 , 7697-37-2 , 7732-18-5
204.38
5359464
thallium
Tl
BKVIYDNLLOSFOA-UHFFFAOYSA-N
CHEBI:30440
[Tl]

Specifications

Thallium ICP Standard
7440-28-0 , 7697-37-2 , 7732-18-5
93.0, 4.76, 0.09
1 mL = 1 mg Tl
BKVIYDNLLOSFOA-UHFFFAOYSA-N
thallium
5359464
Natural Poly Bottle
1,000ppm Tl
97.0, 4.95, 0.1
Tl
[Tl]
204.38
CHEBI:30440
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents