missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFF3 Polyclonal Antibody

Description
A suggested positive control is rat small intestine tissue lysate. PA5-21081 can be used with blocking peptide PEP-1195.
Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. Trefoil peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain and clustered in human chromosome 21q22.3. Trefoil Factor 3 (TFF3) is one of three trefoil peptides secreted by epithelial cells that line mucus membranes in the large and small intestines and lower respiratory tract. TFF3 is mainly active as a disulfide-linked homodimer and may have a role in promoting cell migration in healing processes. It is involved in the maintenance and repair of the intestinal mucosa.
Spécifications
Spécifications
| Antigène | TFF3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.02% sodium azide |
| Gène | TFF3 |
| Numéro d’acquisition du gène | Q03191, Q07654, Q62395 |
| Alias du gène | hITF; hP1.B; Intestinal trefoil factor; ITF; mITF; P1B; Polypeptide P1.B; rITF; rP1.B; Tff3; TFI; Trefoil factor 3; trefoil factor 3 (intestinal); trefoil factor 3, intestinal |
| Symboles de gène | TFF3 |
| Afficher plus de résultats |