missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ ClipperMop™ Mop

For use with wet, dry, or sterile wipers

$365.14 - $443.94

Specifications

Product Type Cleanroom Mop
Sterility Sterile
Includes Head Assembly, 30 to 53 in. Telescoping Handle
Certifications/Compliance ISO Class 3 to 8
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions Price Quantity  
Catalog Number Mfr. No. Dimensions Price Quantity  
18-368-40
View Documents
Texwipe™
TX7102
7 x 4 in. Head Assembly
Each for $443.94
Only null left
 
19-034-961
View Documents
Texwipe™
TX7103
11 x 4 in. Head Assembly
Each for $365.14
Only null left
 
Description

Description

For use with wet, dry, or sterile wipers

  • Compatible with protocols for ISO Class 3 to 8 cleanrooms, sterile and aseptic areas
  • Designed to use standard 9 × 9 in. (23 × 23cm) and 12 × 12 in. (30.5 × 30.5cm) cleanroom wipers—eliminate need to qualify and inventory additional wipers or mop covers
  • Patent-pending mophead utilizes nonabrasive polyethylene clips to securely fasten one or multiple wipers to the head
  • Wiper attachment method does not release particles, fibers, or dirt
  • System can be sterilized by gamma irradiation, chemicals, vaporized hydrogen peroxide (VHP), or ethylene oxide (EtO)

Specifications

Specifications

Cleanroom Mop
Sterile
Head Assembly, 30 to 53 in. Telescoping Handle
ISO Class 3 to 8
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'