Learn More
Applied Biosystems™ Tetrazole Acetonitrile Solution
Each contains 450 mL Tetrazole Acetonitrile Solution.
$1538.00 - $1678.00
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
400606
|
Applied Biosystems™
400606 |
180 mL |
chaque for $1,538.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
401173
|
Applied Biosystems™
401173 |
450 mL |
chaque for $1,678.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.- Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
- Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
- Reagent kits are available for your convenience, and save you time and money.
Spécifications
| Shipped at Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.