missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Tetrazole Acetonitrile Solution

Numéro de catalogue. 400606
Encompass
Modifier l'affichage
Click to view available options
Quantity:
180 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
400606 180 mL
401173 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 400606 Fournisseur Applied Biosystems™ Code fournisseur 400606
Il en reste null

Each contains 450 mL Tetrazole Acetonitrile Solution.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.

Spécifications

Packaging Type Shipped at Room Temperature
Quantity 180 mL

For Research Use Only. Not for use in diagnostic procedures.