Learn More
Applied Biosystems™ Tetrazole Acetonitrile DFS Item
This tetrazole acetonitrile solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.
$1384.00 - $2265.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
4340864 DFS Item
View Documents
Applied Biosystems™
4340864
4 x 180 mL
Each for $1,384.00
Only null left
4340859 DFS Item
View Documents
Applied Biosystems™
4340859
4 x 450 mL
Each for $2,265.00
Only null left
Description
Description
TestedOur high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.
  • Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
  • Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For Research Use Only. Not for use in diagnostic procedures.