Learn More
Applied Biosystems™ Tetrazole Acetonitrile DFS Item
This tetrazole acetonitrile solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.
Supplier:  Applied Biosystems™ 4340859
 View more versions of this product
Catalog No. 4340859
Only null left
Explore available promotions
Description
Description
TestedOur high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.
  • Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
  • Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.
Specifications
Specifications
4 x 450 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.