Learn More
Applied Biosystems™ Tetrazole Acetonitrile
Catalog No. 4340864
Encompass
Change view
Click to view available options
Quantity:
4 x 180 mL
4 x 450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4340864 4 x 180 mL
4340859 4 x 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4340864 Supplier Applied Biosystems™ Supplier No. 4340864
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This tetrazole acetonitrile solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing. oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.
Specifications
Quantity 4 x 180 mL
For Research Use Only. Not for use in diagnostic procedures.