missing translation for 'onlineSavingsMsg'
Learn More

Tetrathiafulvalene 7,7,8,8-tetracyanoquinodimethane salt, ≥97.0% (CHNS), MilliporeSigma™ Supelco™

Catalog No. 111014144
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014144 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014144 Supplier Merck Emd Millipore Supplier No. 905481GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TTF-TCNQ is an organic metal-like charge transfer complex, which is mostly used as an electrode.

Chemical Identifiers

MDL Number MFCD10566936
Synonym 7,7,8,8-Tetracyanoquinodimethane Tetrathiafulvalene salt; TCNQ-TTF; TTF-TCNQ

Specifications

Melting Point 230°C
CAS 40210-84-2
Linear Formula C6H4S4 · C12H4N4
Quantity 1 g
Formula Weight 408.54
Percent Purity ≥97.0% (CHNS)