missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrasodium Pyrophosphate Decahydrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 13472-36-1 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | Na4P2O7 · 10H2 O |
| MDL Number | MFCD00149200 |
| Synonym | Sodium pyrophosphate decahydrate; Sodium pyrophosphate tetrabasic decahydrate; Tetrasodium pyrophosphate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 446.06 |
| Grade | Certified Reference Material |