missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylporphyrin (Chlorin free) 98.0+%, TCI America™

Catalog No. T13591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13591G Supplier TCI America Supplier No. T13591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 917-23-7
Molecular Formula C44H30N4
Molecular Weight (g/mol) 614.75
MDL Number MFCD00011680
InChI Key AQPPOLXYUQPDOD-UHFFFAOYSA-N
Synonym tetraphenylporphyrin, 5,10,15,20-tetraphenylporphyrin, meso-tetraphenylporphine, tetraphenylporphine, 5,10,15,20-tetraphenylporphine, meso-tetraphenylporphyrin, 21h,23h-porphine, 5,10,15,20-tetraphenyl, 5,10,15,20-tetraphenyl-21h,23h-porphine, meso-tetraphenylporphyrine, tetraphenyl porphine
PubChem CID 70186
IUPAC Name 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
SMILES N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Purple-Red
Quantity 1 g
Formula Weight 614.75
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylporphyrin (Chlorin free)