missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylurea 98.0+%, TCI America™

Catalog No. T0158500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0158500G 500 g
T015825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0158500G Supplier TCI America Supplier No. T0158500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 632-22-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00008319
InChI Key AVQQQNCBBIEMEU-UHFFFAOYSA-N
Synonym tetramethylurea, temur, urea, tetramethyl, n,n,n',n'-tetramethylurea, tetramethyluree, tmu, urea, 1,1,3,3-tetramethyl, urea, n,n,n',n'-tetramethyl, tetramethyluree french, tetramethyl-urea
PubChem CID 12437
ChEBI CHEBI:84278
IUPAC Name 1,1,3,3-tetramethylurea
SMILES CN(C)C(=O)N(C)C

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 177°C
Quantity 500 g
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetramethylurea
EINECSNumber (2)-1735
RTECSNumber YU2625000
TSCA Yes