missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane, for the calibration of NMR spectra for NMR spectroscopy, MagniSolv™, MilliporeSigma™

Catalog No. M1081830100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1081830100 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1081830100 Supplier Technidata Supplier No. 1.08183.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For NMR Spectroscopy

Chemical Identifiers

CAS 75-76-3
Molecular Formula C4H12Si
Molecular Weight (g/mol) 88.23
MDL Number MFCD00008274
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
PubChem CID 6396
ChEBI CHEBI:85361
IUPAC Name tetramethylsilane
SMILES C[Si](C)(C)C

Specifications

Physical Form Liquid
Packaging Glass Bottle
Quantity 100 mL
Vapor Pressure 750 hPa at 20°C (68°F)
Odor Characteristic
Formula Weight 88.23
Chemical Name or Material Tetramethylsilane