missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium formate, 30% w/w aq. soln.

Catalog No. p-7052166
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42710AE 100 mL
AA42710AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42710AE Supplier Thermo Scientific Chemicals Supplier No. 042710AE

CAS: 59138-84-0 | C5H13NO2 | 119.16 g/mol

Tetramethylammonium formate may provide the lowest temperature sensitivity, additionally reducing corrosion of stainless-steel flow system by chloride-induced depassivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59138-84-0
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00054402
InChI Key WWIYWFVQZQOECA-UHFFFAOYSA-M
Synonym tetramethylammonium formate, fibrogenina, forgenin, tonoformina, methanaminium, n,n,n-trimethyl-, formate, tetramethylazanium formate, n,n,n-trimethylmethanaminium formate, ammonium, tetramethyl-, formate, methanaminium, n,n,n-trimethyl-, formate 1:1, c4h12n.cho2
PubChem CID 42961
IUPAC Name tetramethylazanium;formate
SMILES [O-]C=O.C[N+](C)(C)C

Specifications

Linear Formula (CH3)4N(COOH)
Quantity 100 mL
Beilstein 3916177
Solubility Information Fully miscible in water.
Formula Weight 119.16
Concentration or Composition (by Analyte or Components) 30% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetramethylammonium formate
EINECSNumber 261-622-8
RTECSNumber BS7960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only