Learn More
Tetramethyl orthocarbonate, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 1850-14-2 |
|---|---|
| Molecular Formula | C5H12O4 |
| Molecular Weight (g/mol) | 136.15 |
| InChI Key | AHJWSRRHTXRLAQ-UHFFFAOYSA-N |
| Synonym | tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc |
| PubChem CID | 74613 |
| IUPAC Name | tetramethoxymethane |
| SMILES | COC(OC)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC158110050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC158110250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1850-14-2 | |
| 136.15 | |
| tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc | |
| tetramethoxymethane |
| C5H12O4 | |
| AHJWSRRHTXRLAQ-UHFFFAOYSA-N | |
| 74613 | |
| COC(OC)(OC)OC |
Specifications
| 1850-14-2 | |
| 114.0°C | |
| Authentic | |
| C5H12O4 | |
| C(OCH3)4 | |
| 03,285 | |
| tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc | |
| COC(OC)(OC)OC | |
| 136.15 | |
| 136.15 | |
| Tetramethyl orthocarbonate |
| 1.0200g/mL | |
| 22°C | |
| 98.5% min. (GC) | |
| 1.3845 | |
| 03,II,4 | |
| 1.02 | |
| AHJWSRRHTXRLAQ-UHFFFAOYSA-N | |
| tetramethoxymethane | |
| 74613 | |
| 99% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 217-438-5
RUO – Research Use Only