missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethrin, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC S4251
Comprend: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Tetramethrin | 7696-12-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
| Tetramethrin | |
| 7696-12-0,67-56-1 | |
| 99.9,0.1 | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| CXBMCYHAMVGWJQ-UHFFFAOYSA-N | |
| Methanol-P and T | |
| 331.412 | |
| 83975 | |
| 1000μg/mL |
| 2mL amber prescored glass ampule | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| C19H25NO4 | |
| Certified Reference Material | |
| CC(=CC1C(C1(C)C)C(=O)OCN2C(=O)C3=C(C2=O)CCCC3)C | |
| (1,3-dioxo-4,5,6,7-tetrahydroisoindol-2-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate | |
| 1 mL | |
| CHEBI:39397 |
Identifiants chimiques
| 7696-12-0 | |
| 331.412 | |
| 83975 | |
| (1,3-dioxo-4,5,6,7-tetrahydroisoindol-2-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate |
| C19H25NO4 | |
| CXBMCYHAMVGWJQ-UHFFFAOYSA-N | |
| CHEBI:39397 | |
| CC(=CC1C(C1(C)C)C(=O)OCN2C(=O)C3=C(C2=O)CCCC3)C |
Sécurité et manipulation
Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Freezer
Product intended for laboratory use only