missing translation for 'onlineSavingsMsg'
Learn More

Tetramethoxymethane 97.0+%, TCI America™

Catalog No. T10455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10455G 5 g
T104525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10455G Supplier TCI America Supplier No. T10455G

Chemical Identifiers

CAS 1850-14-2
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00008473
InChI Key AHJWSRRHTXRLAQ-UHFFFAOYSA-N
Synonym tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
PubChem CID 74613
IUPAC Name tetramethoxymethane
SMILES COC(OC)(OC)OC

Specifications

Color Colorless
Boiling Point 114°C
Quantity 5 g
UN Number 3272
Formula Weight 136.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetramethoxymethane