missing translation for 'onlineSavingsMsg'
Learn More

beta-Tetralone, 95%

Catalog No. AC302830250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302830250 25 g Glass bottle
AC302830050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC302830250 Supplier Thermo Scientific Chemicals Supplier No. 302830250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 530-93-8 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 530-93-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001727
InChI Key KCKZIWSINLBROE-UHFFFAOYSA-N
Synonym 2-tetralone, beta-tetralone, 1,2,3,4-tetrahydro-2-naphthalenone, 3,4-dihydronaphthalen-2 1h-one, 1,2,3,4-tetrahydronaphthalen-2-one, 2 1h-naphthalenone, 3,4-dihydro, .beta.-tetralone, tetralin-2-one, 3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-2-oxonaphthalene
PubChem CID 68266
IUPAC Name 3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=CC=CC=C2CC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 18.0°C
Color Yellow
Density 1.1060g/mL
Boiling Point 131.0°C to 133.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5590 to 1.5630
Quantity 25 g
Beilstein 07, 370
Specific Gravity 1.106
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and benzene
Formula Weight 146.19
Percent Purity 95%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material β-Tetralone, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu
EINECSNumber 208-498-3
TSCA TSCA

RUO – Research Use Only