missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(methylthio)tetrathiafulvalene 96.0+%, TCI America™

Catalog No. T1119100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1119100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1119100MG Supplier TCI America Supplier No. T1119100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51501-77-0
Molecular Formula C10H12S8
Molecular Weight (g/mol) 388.686
MDL Number MFCD00059747
InChI Key LLLXUARUONPZIY-UHFFFAOYSA-N
Synonym TMT-TTF
PubChem CID 633638
IUPAC Name 2-[4,5-bis(methylsulfanyl)-1,3-dithiol-2-ylidene]-4,5-bis(methylsulfanyl)-1,3-dithiole
SMILES CSC1=C(SC(=C2SC(=C(S2)SC)SC)S1)SC

Specifications

Melting Point 96°C
Quantity 100 mg
Formula Weight 388.69
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(methylthio)tetrathiafulvalene [Organic Electronic Material]