missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)diboron, Thermo Scientific™

Catalog No. AC468540050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468540050 5 g
AC468540250 25 g
AC468541000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC468540050 Supplier Thermo Scientific Chemicals Supplier No. 468540050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1630-79-1
Molecular Formula C8H24B2N4
Molecular Weight (g/mol) 197.93
MDL Number MFCD00048012
InChI Key KMCDRSZVZMXKRL-UHFFFAOYSA-N
IUPAC Name dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine
SMILES CN(C)B(B(N(C)C)N(C)C)N(C)C

Specifications

Melting Point -33°C
Density 0.926
Boiling Point 55°C to 57°C (2.5 mmHg)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index 1.4680 to 1.4760 (20°C, 589 nm)
Quantity 5 g
Formula Weight 197.93
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)diboron
Hazard Category GHS07: Exclamation mark
Hazard Statement GHS08: Health hazard
ShelfLife 2 years
Recommended Storage Refrigerator +4°C