missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2,4-pentanedionato)zirconium(IV), TCI America™

Catalog No. Z000625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z000625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z000625G Supplier TCI America Supplier No. Z000625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17501-44-9
Molecular Formula C20H28O8Zr
Molecular Weight (g/mol) 487.66
MDL Number MFCD00000036
InChI Key DFPGFWYWSAYLCW-UHFFFAOYSA-N
Synonym 4-hydroxypent-3-en-2-one; zirconium
PubChem CID 50912253
IUPAC Name zirconium(4+) tetrakis(2,4-dioxopentan-3-ide)
SMILES [Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 192°C
Color White-Yellow
pH 7.2 (4.5g/L 20°C)
Quantity 25 g
Formula Weight 487.66
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(2,4-pentanedionato)zirconium(IV)
EINECSNumber (2)-2128&(2)-2193&(2)-2142
RTECSNumber ZH9280000
TSCA Yes